Assay Details
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252580.1 | NP_001239509.1 | 10 - 11 | 1476 | 66 | |
NM_019729.3 | NP_062703.2 | 10 - 11 | 1443 | 66 | ||
GenBank mRNA | AB045709.1 | - | 9 - 10 | 1217 | 66 | |
AF057146.1 | - | 9 - 10 | 1234 | 66 | ||
AK122195.1 | - | 10 - 11 | 1354 | 66 | ||
AK129003.1 | - | 10 - 11 | 1444 | 66 | ||
AK134058.1 | - | 10 - 11 | 1398 | 66 | ||
AK142881.1 | - | 7 - 8 | 941 | 66 | ||
AK153015.1 | - | 9 - 10 | 1191 | 66 | ||
AK163469.1 | - | 10 - 11 | 1376 | 66 | ||
BC038123.1 | - | 10 - 11 | 1428 | 66 | ||
BC050947.1 | - | 10 - 11 | 1408 | 6825981 | 66 | |
BC061465.1 | - | 10 - 11 | 1408 | 6825981 | 66 | |
BC066126.1 | - | 10 - 11 | 1406 | 30301665 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
