Assay Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 11 - 12 | 1860 | 93 | |
NM_019867.2 | NP_063920.2 | 9 - 10 | 1497 | 93 | ||
GenBank mRNA | AJ238898.1 | - | 9 - 10 | 1478 | 93 | |
AK138433.1 | - | 9 - 10 | 1466 | 93 | ||
AY038025.1 | - | 9 - 10 | 1472 | 93 | ||
BC022680.1 | - | 3 - 4 | 275 | 93 | ||
BC039279.1 | - | 11 - 12 | 1860 | 4973826 | 93 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC022680.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
