Assay Details
Gene Symbol: Rps6ka4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019924.2 | NP_064308.2 | 4 - 5 | 543 | 71 | |
GenBank mRNA | AF074714.1 | - | 4 - 5 | 515 | 71 | |
AK089467.1 | - | 4 - 5 | 534 | 71 | ||
AK154674.1 | - | 4 - 5 | 542 | 71 | ||
BC012964.1 | - | 4 - 5 | 553 | 4160873 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
