Assay Details
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135727.2 | NP_001129199.1 | 6 - 7 | 1137 | 80 | |
NM_021389.6 | NP_067364.2 | 5 - 6 | 702 | 80 | ||
NR_110969.1 | 5 - 6 | 702 | 80 | |||
NR_110970.1 | 5 - 6 | 702 | 80 | |||
GenBank mRNA | AK004636.1 | - | 5 - 6 | 702 | 80 | |
AK018032.1 | - | 5 - 6 | 718 | 80 | ||
AK020782.1 | - | 5 - 6 | 679 | 80 | ||
AK030839.1 | - | 5 - 6 | 703 | 80 | ||
AK038540.1 | - | 8 - 9 | 1001 | 80 | ||
AK049182.1 | - | 6 - 7 | 1138 | 80 | ||
AK080930.1 | - | 6 - 7 | 1002 | 80 | ||
AK172022.1 | - | 6 - 7 | 1118 | 80 | ||
BC034646.1 | - | 5 - 6 | 706 | 80 | ||
BC052863.1 | - | 5 - 6 | 651 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:

Citations (1)
Delineating biological pathways unique to embryonic stem cell-derived insulin-producing cell lines from their noninsulin-producing progenitor cell lines.
- Authors
- Chen TS, Tan SS, Yeo RW, Teh BJ, Luo R, Li G, Lim SK
- Journal
- Endocrinology - Page No 3600 (Sun Aug 01 00:00:00 UTC 2010)
- Volume (Year)
- 151 (2010)
- Product Usage
- -
