Assay Details
Gene Symbol: Nfatc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168346.1 | NP_001161818.1 | 1 - 2 | 349 | 99 | |
NM_023699.3 | NP_076188.3 | 1 - 2 | 349 | 99 | ||
GenBank mRNA | AK159078.1 | - | 1 - 2 | 195 | 99 | |
BC028928.1 | - | 1 - 2 | 136 | 3670623 | 99 | |
CJ067971.1 | - | 1 - 2 | 350 | 99 | ||
EU887656.1 | - | 1 - 2 | 107 | 99 | ||
EU887657.1 | - | 1 - 2 | 107 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
