Assay Details
Gene Symbol: Scyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023912.2 | NP_076401.1 | 3 - 4 | 673 | 53 | |
GenBank mRNA | AF276514.1 | - | 3 - 4 | 673 | 53 | |
AK019240.1 | - | 3 - 4 | 666 | 53 | ||
AK134356.1 | - | 3 - 4 | 672 | 53 | ||
AK144134.1 | - | 3 - 4 | 659 | 53 | ||
AK145869.1 | - | 3 - 4 | 693 | 53 | ||
AK161813.1 | - | 3 - 4 | 672 | 53 | ||
AK181752.1 | - | 1 - 2 | 43 | 53 | ||
BC034519.1 | - | 3 - 4 | 659 | 3672524 | 53 | |
HM856325.1 | - | 3 - 4 | 673 | 53 | ||
HM856326.1 | - | 3 - 4 | 673 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
