Assay Details
Gene Symbol: Usp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301628.1 | NP_001288557.1 | 7 - 8 | 931 | 81 | |
NM_027604.4 | NP_081880.2 | 8 - 9 | 1018 | 81 | ||
GenBank mRNA | AF468037.1 | - | 8 - 9 | 922 | 81 | |
AK014891.1 | - | 2 - 3 | 229 | 81 | ||
AK052398.1 | - | 6 - 7 | 747 | 81 | ||
AK122301.1 | - | 7 - 8 | 846 | 81 | ||
AK133852.1 | - | 7 - 8 | 840 | 81 | ||
AK145749.1 | - | 8 - 9 | 922 | 81 | ||
AK161957.1 | - | 7 - 8 | 879 | 81 | ||
AK163315.1 | - | 7 - 8 | 860 | 81 | ||
AK168755.1 | - | 7 - 8 | 866 | 81 | ||
BC050042.1 | - | 8 - 9 | 934 | 30022307 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
