Assay Details
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122829.1 | NP_001116301.1 | 21 - 22 | 3318 | 77 | |
NM_030680.2 | NP_109605.2 | 21 - 22 | 3285 | 77 | ||
GenBank mRNA | AF182947.1 | - | 22 - 23 | 77 | ||
AF322655.1 | - | 21 - 22 | 3005 | 77 | ||
AK052517.1 | - | 10 - 11 | 1353 | 77 | ||
AK148196.1 | - | 22 - 23 | 3295 | 77 | ||
BC030916.1 | - | 9 - 10 | 1296 | 77 | ||
BC052149.1 | - | 21 - 22 | 3223 | 6332288 | 77 | |
BC056442.1 | - | 21 - 22 | 3318 | 5702493 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
