Assay Details
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031186.3 | NP_112463.2 | 1 - 2 | 1141 | 61 | |
NR_121616.1 | 2 - 3 | 1289 | 61 | |||
GenBank mRNA | AF221095.1 | - | 1 - 2 | 1140 | 61 | |
AK141945.1 | - | 2 - 3 | 1290 | 61 | ||
BC079622.1 | - | 1 - 2 | 1056 | 30535990 | 61 | |
BC112404.1 | - | 1 - 2 | 1053 | 40060424 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
