Assay Details
Gene Symbol: Sardh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138665.2 | NP_619606.1 | 12 - 13 | 2229 | 63 | |
GenBank mRNA | AK087163.1 | - | 5 - 6 | 530 | 63 | |
AK140451.1 | - | 11 - 12 | 1505 | 63 | ||
AK159236.1 | - | 12 - 13 | 2193 | 63 | ||
AK159681.1 | - | 12 - 13 | 2193 | 63 | ||
AK159704.1 | - | 12 - 13 | 2193 | 63 | ||
AK163515.1 | - | 12 - 13 | 1758 | 63 | ||
AK163660.1 | - | 11 - 12 | 1505 | 63 | ||
BC003456.1 | - | 12 - 13 | 1738 | 2648344 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
