Assay Details
Gene Symbol: Tmlhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138758.1 | NP_620097.1 | 2 - 3 | 310 | 70 | |
GenBank mRNA | AK051339.1 | - | 2 - 3 | 303 | 70 | |
AK052425.1 | - | 2 - 3 | 312 | 70 | ||
AK144627.1 | - | 2 - 3 | 306 | 70 | ||
AK161724.1 | - | 2 - 3 | 277 | 70 | ||
AY033513.1 | - | 1 - 2 | 148 | 70 | ||
BC010495.1 | - | 2 - 3 | 310 | 4195680 | 70 | |
BC115365.1 | - | 2 - 3 | 242 | 40059106 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
