Assay Details
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012363.2 | NP_001012363.2 | 1 - 2 | 71 | 55 | |
NM_145559.2 | NP_663534.1 | 2 - 3 | 265 | 55 | ||
GenBank mRNA | AY776155.1 | - | 1 - 2 | 50 | 55 | |
BC006076.1 | - | 2 - 3 | 265 | 3593581 | 55 | |
BC138213.1 | - | 2 - 3 | 206 | 8861232 | 55 | |
BC145198.1 | - | 2 - 3 | 206 | 9053749 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
