Assay Details
Gene Symbol: Usp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145628.4 | NP_663603.3 | 17 - 18 | 2340 | 58 | |
GenBank mRNA | AK031767.1 | - | 17 - 18 | 2319 | 58 | |
AK169116.1 | - | 9 - 10 | 1163 | 58 | ||
AK172230.1 | - | 4 - 5 | 1303 | 58 | ||
AK220478.1 | - | 15 - 16 | 2855 | 58 | ||
BC005470.1 | - | 15 - 16 | 1909 | 3496607 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
