Assay Details
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 10 - 11 | 1354 | 94 | |
GenBank mRNA | BC031778.1 | - | 901 | 4501115 | 94 | |
BC049904.1 | - | 901 | 4501115 | 94 | ||
BC064815.1 | - | 10 - 11 | 1339 | 6516981 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
