Assay Details
Gene Symbol: Pknox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148950.3 | NP_683752.2 | 2 - 3 | 147 | 111 | |
GenBank mRNA | AF487460.1 | - | 1 - 2 | 53 | 111 | |
BC050865.1 | - | 2 - 3 | 81 | 6332968 | 111 | |
BY346714.1 | - | 2 - 3 | 92 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF487460.1, BC050865.1, NM_148950.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
