Assay Details
Gene Symbol: Blnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008528.4 | NP_032554.2 | 2 - 3 | 592 | 105 | |
GenBank mRNA | AB015290.1 | - | 2 - 3 | 266 | 105 | |
AF068182.1 | - | 2 - 3 | 247 | 105 | ||
AJ222814.1 | - | 2 - 3 | 217 | 105 | ||
AK089704.1 | - | 2 - 3 | 232 | 105 | ||
AK172016.1 | - | 2 - 3 | 219 | 105 | ||
BC059785.1 | - | 2 - 3 | 218 | 5339723 | 105 | |
Y17159.1 | - | 2 - 3 | 592 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
