Assay Details
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008746.5 | NP_032772.3 | 8 - 9 | 1255 | 68 | |
NM_182809.2 | NP_877961.1 | 8 - 9 | 1255 | 68 | ||
GenBank mRNA | AF035399.1 | - | 9 - 10 | 1350 | 68 | |
AF035400.1 | - | 8 - 9 | 1404 | 68 | ||
AY336094.1 | - | 8 - 9 | 1200 | 68 | ||
BC139764.1 | - | 9 - 10 | 1267 | 40110345 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
