Assay Details
Gene Symbol: Ntrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008746.5 | NP_032772.3 | 9 - 10 | 1287 | 71 | |
NM_182809.2 | NP_877961.1 | 9 - 10 | 1287 | 71 | ||
GenBank mRNA | AF035399.1 | - | 10 - 11 | 1382 | 71 | |
AF035400.1 | - | 9 - 10 | 1436 | 71 | ||
AY336094.1 | - | 9 - 10 | 1232 | 71 | ||
BC139764.1 | - | 10 - 11 | 1299 | 40110345 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
