Assay Details
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290448.1 | NP_001277377.1 | 8 - 9 | 1796 | 82 | |
NM_001290449.1 | NP_001277378.1 | 8 - 9 | 1796 | 82 | ||
NM_023739.3 | NP_076228.2 | 8 - 9 | 1759 | 82 | ||
GenBank mRNA | AF223576.1 | - | 8 - 9 | 1723 | 82 | |
AK005038.2 | - | 8 - 9 | 1731 | 82 | ||
AK033850.1 | - | 8 - 9 | 1690 | 82 | ||
AK050749.1 | - | 8 - 9 | 1733 | 82 | ||
AK144647.1 | - | 8 - 9 | 1745 | 82 | ||
AK146108.1 | - | 8 - 9 | 1713 | 82 | ||
AK155387.1 | - | 8 - 9 | 1711 | 82 | ||
AK160013.1 | - | 8 - 9 | 1729 | 82 | ||
BC053709.1 | - | 8 - 9 | 1694 | 30052746 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
