Assay Details
Gene Symbol: Wwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025830.3 | NP_080106.1 | 1 - 2 | 47 | 80 | |
GenBank mRNA | AK004962.2 | - | 1 - 2 | 63 | 80 | |
AK141281.1 | - | 1 - 2 | 57 | 80 | ||
BC048184.1 | - | 1 - 2 | 47 | 6827381 | 80 | |
JN712751.1 | - | 1 - 2 | 80 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK004962.2, BC048184.1, JN712751.1, NM_025830.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
