Assay Details
Gene Symbol: Wwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025830.3 | NP_080106.1 | 21 - 22 | 2401 | 64 | |
GenBank mRNA | AK004452.1 | - | 1 - 2 | 58 | 64 | |
AK004962.2 | - | 21 - 22 | 2417 | 64 | ||
AK088936.1 | - | 21 - 22 | 2380 | 64 | ||
AK090392.1 | - | 251 | 64 | |||
AK146306.1 | - | 20 - 21 | 2263 | 64 | ||
AK151548.1 | - | 21 - 22 | 2395 | 64 | ||
AK159248.1 | - | 12 - 13 | 1445 | 64 | ||
BC004712.1 | - | 6 - 7 | 721 | 64 | ||
BC018496.1 | - | 5 - 6 | 1599 | 64 | ||
BC039921.1 | - | 21 - 22 | 2393 | 4954925 | 64 | |
BC048184.1 | - | 21 - 22 | 2401 | 6827381 | 64 | |
HQ896246.1 | - | 12 - 13 | 1498 | 64 | ||
JN712751.1 | - | 21 - 22 | 2434 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
