Assay Details
Gene Symbol: Krit1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170552.1 | NP_001164023.1 | 5 - 6 | 857 | 98 | |
NM_030675.3 | NP_109600.2 | 5 - 6 | 811 | 98 | ||
NR_033173.1 | 5 - 6 | 910 | 98 | |||
GenBank mRNA | AF306509.1 | - | 5 - 6 | 524 | 98 | |
AF310134.1 | - | 5 - 6 | 787 | 98 | ||
AK039264.1 | - | 832 | 98 | |||
AK144667.1 | - | 5 - 6 | 793 | 98 | ||
AY328895.1 | - | 4 - 5 | 493 | 98 | ||
AY464945.1 | - | 4 - 5 | 493 | 98 | ||
BC016087.1 | - | 5 - 6 | 761 | 98 | ||
BC044820.1 | - | 4 - 5 | 552 | 98 | ||
BC054819.1 | - | 5 - 6 | 902 | 98 | ||
BC085252.1 | - | 5 - 6 | 617 | 98 | ||
BC141042.1 | - | 5 - 6 | 655 | 9055654 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
