Assay Details
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164635.1 | NP_001158107.1 | 1 - 2 | 109 | 58 | |
NM_031194.5 | NP_112471.3 | 2 - 3 | 281 | 58 | ||
GenBank mRNA | AF078869.1 | - | 1 - 2 | 56 | 58 | |
AK031962.1 | - | 2 - 3 | 247 | 58 | ||
AK044336.1 | - | 1 - 2 | 78 | 58 | ||
AK157891.1 | - | 1 - 2 | 110 | 58 | ||
BB622154.1 | - | 2 - 3 | 244 | 58 | ||
BB843926.1 | - | 1 - 2 | 110 | 58 | ||
BC014762.1 | - | 1 - 2 | 87 | 4239544 | 58 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF078869.1,AK031962.1,AK044336.1,AK157891.1,BC014762.1,NM_001164635.1,NM_031194.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
