Assay Details
Gene Symbol: Parvb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133167.3 | NP_573395.1 | 4 - 5 | 408 | 80 | |
GenBank mRNA | AF237770.1 | - | 4 - 5 | 386 | 80 | |
AK143614.1 | - | 4 - 5 | 388 | 80 | ||
AK147758.1 | - | 4 - 5 | 388 | 80 | ||
BC138747.1 | - | 4 - 5 | 468 | 8861769 | 80 | |
BC138748.1 | - | 4 - 5 | 468 | 8861770 | 80 | |
BC158113.1 | - | 4 - 5 | 607 | 9088187 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
