Assay Details
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 1 - 2 | 253 | 97 | |
GenBank mRNA | AJ344447.1 | - | 1 - 2 | 253 | 97 | |
AK047460.1 | - | 1 - 2 | 184 | 97 | ||
AK171707.1 | - | 1 - 2 | 177 | 97 | ||
BC066173.1 | - | 1 - 2 | 206 | 30049516 | 97 | |
DV651307.1 | - | 1 - 2 | 283 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
