Assay Details
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133891.3 | NP_598652.3 | 12 - 13 | 1769 | 76 | |
GenBank mRNA | BC025941.1 | - | 3 - 4 | 331 | 76 | |
BC113167.1 | - | 12 - 13 | 1525 | 40059854 | 76 | |
BC113169.1 | - | 12 - 13 | 1525 | 40059860 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
