Assay Details
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144803.2 | NP_659052.1 | 1 - 2 | 48 | 58 | |
GenBank mRNA | BC011490.1 | - | 1 - 2 | 48 | 4193582 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC011490.1, NM_144803.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
