Assay Details
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153171.4 | NP_694811.1 | 4 - 5 | 315 | 131 | |
GenBank mRNA | AF498319.1 | - | 4 - 5 | 364 | 131 | |
AK043268.1 | - | 4 - 5 | 315 | 131 | ||
BC049624.1 | - | 4 - 5 | 233 | 6585574 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
