Assay Details
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153543.2 | NP_705771.2 | 9 - 10 | 1264 | 64 | |
GenBank mRNA | AK052361.1 | - | 9 - 10 | 1155 | 64 | |
AK086413.1 | - | 9 - 10 | 1208 | 64 | ||
AK141954.1 | - | 9 - 10 | 1226 | 64 | ||
AK208193.1 | - | 2 - 3 | 153 | 64 | ||
BC034531.1 | - | 9 - 10 | 1186 | 4188055 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
