Assay Details
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153551.1 | NP_705779.1 | 2 - 3 | 187 | 53 | |
GenBank mRNA | AK014482.1 | - | 2 - 3 | 192 | 53 | |
AK040537.1 | - | 2 - 3 | 117 | 53 | ||
AK170891.1 | - | 2 - 3 | 185 | 53 | ||
BC035208.1 | - | 2 - 3 | 187 | 3671384 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
