Assay Details
Gene Symbol: Thoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153552.3 | NP_705780.1 | 19 - 20 | 1608 | 106 | |
GenBank mRNA | AK031785.1 | - | 19 - 20 | 1609 | 106 | |
AK032200.1 | - | 19 - 20 | 1609 | 106 | ||
AK052835.1 | - | 19 - 20 | 1604 | 106 | ||
AK167374.1 | - | 19 - 20 | 1622 | 106 | ||
AK168619.1 | - | 19 - 20 | 1874 | 106 | ||
BC024951.1 | - | 19 - 20 | 1780 | 4010605 | 106 | |
BC027149.1 | - | 12 - 13 | 1387 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
