Assay Details
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153585.5 | NP_705813.2 | 3 - 4 | 540 | 68 | |
GenBank mRNA | AK029103.1 | - | 3 - 4 | 415 | 68 | |
AK080503.1 | - | 3 - 4 | 427 | 68 | ||
AK149944.1 | - | 3 - 4 | 373 | 68 | ||
AK157824.1 | - | 3 - 4 | 439 | 68 | ||
AK161842.1 | - | 3 - 4 | 524 | 68 | ||
AK171701.1 | - | 3 - 4 | 520 | 68 | ||
BC025554.1 | - | 3 - 4 | 401 | 5321869 | 68 | |
BC036179.1 | - | 3 - 4 | 512 | 5059723 | 68 | |
BC039183.1 | - | 3 - 4 | 514 | 2631793 | 68 | |
CA537067.1 | - | 3 - 4 | 540 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
