Assay Details
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153680.2 | NP_710147.1 | 3 - 4 | 357 | 90 | |
GenBank mRNA | AK047265.1 | - | 3 - 4 | 357 | 90 | |
AK077650.1 | - | 3 - 4 | 359 | 90 | ||
AK077780.1 | - | 3 - 4 | 359 | 90 | ||
AK156299.1 | - | 3 - 4 | 408 | 90 | ||
BC023732.1 | - | 3 - 4 | 373 | 5352355 | 90 | |
BC026571.1 | - | 3 - 4 | 357 | 4986268 | 90 | |
BC029675.1 | - | 3 - 4 | 344 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
