Assay Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009481.2 | NP_033507.2 | 38 - 39 | 6961 | 88 | |
GenBank mRNA | AK034941.1 | - | 4 - 5 | 1022 | 88 | |
DQ086491.1 | - | 38 - 39 | 6922 | 88 | ||
U67874.1 | - | 38 - 39 | 6922 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
