Assay Details
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 8 - 9 | 794 | 93 | |
GenBank mRNA | AB007544.1 | - | 8 - 9 | 771 | 93 | |
AB011543.1 | - | 8 - 9 | 783 | 93 | ||
AK084827.1 | - | 8 - 9 | 795 | 93 | ||
D87521.1 | - | 8 - 9 | 771 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
