Assay Details
Gene Symbol: Prkdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011159.2 | NP_035289.2 | 68 - 69 | 9582 | 88 | |
GenBank mRNA | AB007544.1 | - | 68 - 69 | 9559 | 88 | |
AB011543.1 | - | 68 - 69 | 9571 | 88 | ||
D87521.1 | - | 68 - 69 | 9559 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
