Assay Details
Gene Symbol: Prpf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133701.2 | NP_598462.1 | 12 - 13 | 1786 | 62 | |
GenBank mRNA | AK081998.1 | - | 12 - 13 | 1721 | 62 | |
AK145461.1 | - | 12 - 13 | 1789 | 62 | ||
AK166890.1 | - | 12 - 13 | 1738 | 62 | ||
AK182198.1 | - | 1 - 2 | 83 | 62 | ||
AK210126.1 | - | 1 - 2 | 83 | 62 | ||
BC005801.1 | - | 2 - 3 | 302 | 62 | ||
BC014869.1 | - | 12 - 13 | 1713 | 3602158 | 62 | |
BC023691.1 | - | 12 - 13 | 1715 | 5344332 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
