Assay Details
Gene Symbol: Mycbpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170671.2 | NP_733771.2 | 13 - 14 | 1810 | 79 | |
GenBank mRNA | AK029929.1 | - | 12 - 13 | 1986 | 79 | |
AK166780.1 | - | 6 - 7 | 958 | 79 | ||
BC027327.1 | - | 9 - 10 | 1171 | 3991131 | 79 | |
BC050808.1 | - | 2 - 3 | 333 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
