Assay Details
Gene Symbol: Hgs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159328.1 | NP_001152800.1 | 2 - 3 | 214 | 101 | |
NM_008244.3 | NP_032270.3 | 2 - 3 | 214 | 101 | ||
GenBank mRNA | AK046299.1 | - | 2 - 3 | 175 | 101 | |
AK145031.1 | - | 2 - 3 | 134 | 101 | ||
AK166441.1 | - | 2 - 3 | 188 | 101 | ||
BC003239.1 | - | 2 - 3 | 158 | 3497966 | 101 | |
D50050.1 | - | 2 - 3 | 164 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
