Assay Details
Gene Symbol: Hipk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010434.2 | NP_034564.2 | 9 - 10 | 2555 | 143 | |
GenBank mRNA | AF077660.1 | - | 9 - 10 | 2309 | 143 | |
AF170305.1 | - | 8 - 9 | 2019 | 143 | ||
AF305238.1 | - | 8 - 9 | 2017 | 143 | ||
AK138905.1 | - | 1 - 2 | 2965 | 143 | ||
AK140130.1 | - | 9 - 10 | 2658 | 143 | ||
BC131663.1 | - | 9 - 10 | 2187 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
