Assay Details
Gene Symbol: Ecel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277925.1 | NP_001264854.1 | 2 - 3 | 1000 | 108 | |
NM_021306.3 | NP_067281.2 | 2 - 3 | 992 | 108 | ||
GenBank mRNA | AB026294.1 | - | 1 - 2 | 820 | 108 | |
AK039467.1 | - | 2 - 3 | 992 | 108 | ||
AK159365.1 | - | 2 - 3 | 1002 | 108 | ||
BC057569.1 | - | 1 - 2 | 822 | 6413731 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
