Assay Details
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113573.1 | NP_001107045.1 | 6 - 7 | 1277 | 68 | |
NM_001113574.1 | NP_001107046.1 | 6 - 7 | 1277 | 68 | ||
NM_023336.4 | NP_075825.3 | 6 - 7 | 1437 | 68 | ||
GenBank mRNA | AB206708.2 | - | 5 - 6 | 1088 | 68 | |
AF269193.1 | - | 6 - 7 | 1225 | 68 | ||
AK037435.1 | - | 6 - 7 | 1259 | 68 | ||
AK076472.1 | - | 6 - 7 | 1208 | 68 | ||
AK076924.1 | - | 6 - 7 | 1356 | 68 | ||
AK081471.1 | - | 6 - 7 | 1258 | 68 | ||
AK136007.1 | - | 6 - 7 | 1237 | 68 | ||
AK136429.1 | - | 6 - 7 | 1208 | 68 | ||
AK159827.1 | - | 6 - 7 | 1263 | 68 | ||
AK160752.1 | - | 6 - 7 | 1236 | 68 | ||
AK161164.1 | - | 6 - 7 | 1233 | 68 | ||
AK167573.1 | - | 6 - 7 | 1235 | 68 | ||
AY513269.1 | - | 5 - 6 | 1103 | 68 | ||
BC004832.1 | - | 6 - 7 | 1222 | 68 | ||
BC031536.1 | - | 6 - 7 | 1251 | 4015879 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
