Assay Details
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023397.4 | NP_075886.1 | 1 - 2 | 173 | 69 | |
NR_028316.1 | 1 - 2 | 173 | 69 | |||
GenBank mRNA | AF230273.1 | - | 1 - 2 | 39 | 69 | |
AK007319.1 | - | 1 - 2 | 157 | 69 | ||
AK007694.1 | - | 1 - 2 | 117 | 69 | ||
AK008881.1 | - | 1 - 2 | 88 | 69 | ||
AK160438.1 | - | 1 - 2 | 103 | 69 | ||
BC046613.1 | - | 1 - 2 | 96 | 6465642 | 69 | |
CJ176407.1 | - | 1 - 2 | 174 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK007694.1, AK008881.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
