Assay Details
Gene Symbol: Mdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023397.4 | NP_075886.1 | 5 - 6 | 541 | 145 | |
NR_028316.1 | 4 - 5 | 610 | 145 | |||
GenBank mRNA | AF230273.1 | - | 5 - 6 | 407 | 145 | |
AK007319.1 | - | 5 - 6 | 525 | 145 | ||
AK007694.1 | - | 4 - 5 | 554 | 145 | ||
AK008881.1 | - | 4 - 5 | 525 | 145 | ||
AK160438.1 | - | 5 - 6 | 471 | 145 | ||
BC046613.1 | - | 5 - 6 | 464 | 6465642 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
dephosphorylation
fructosamine metabolic process
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
metal ion binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
protein histidine phosphatase activity
inosine-diphosphatase activity
dephosphorylation
fructosamine metabolic process
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
JUN kinase phosphatase activity
inositol bisphosphate phosphatase activity
hydrolase activity
phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
NADP phosphatase activity
transmembrane receptor protein phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 3-phosphatase activity
inositol-1,4,5,6-tetrakisphosphate 6-phosphatase activity
inositol-4,5-bisphosphate 5-phosphatase activity
5-amino-6-(5-phosphoribitylamino)uracil phosphatase activity
phosphatidylinositol-4-phosphate phosphatase activity
phosphatidylinositol-3,5-bisphosphate 5-phosphatase activity
metal ion binding
phosphatidylinositol-3,5-bisphosphate 3-phosphatase activity
inositol phosphate phosphatase activity
inositol-1,3,4,5,6-pentakisphosphate 1-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 1-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 6-phosphatase activity
inositol-1,3,4,6-tetrakisphosphate 1-phosphatase activity
phosphatidylinositol-1,4,5-trisphosphate 5-phosphatase activity
protein histidine phosphatase activity
inosine-diphosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619153 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619152 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619151 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | BC046613.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | AK007694.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | BC046613.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P139740 | AK007694.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 694376 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149006 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | BC046613.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | AK007694.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | BC046613.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P139740 | AK007694.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619148 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619136 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619144 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29619137 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149006 | - | - |
Whole Mouse Genome G4122A | A_51_P102652 | AK008881.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102652 | AK160438.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102652 | BC046613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102652 | NR_028316.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P102652 | AF230273.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | BC046613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | AK007694.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | BC046613.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | NR_028316.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | NM_023397.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P139740 | AK007694.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117134 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149006 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11787155 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11333400 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11333409 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11333405 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11333403 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11333410 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619152 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619151 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P29619153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29619146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619120 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619132 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11434377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11262733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619125 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619128 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619124 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619119 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619122 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619130 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619152 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619136 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619116 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29619156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10420086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420083 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420087 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420088 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420085 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10420084 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450067_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428337_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10420080 | - | - |
Mouse Genome 430 2.0 Array | 1428337_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420974_at | - | - |
Mouse Genome 430 2.0 Array | 1450067_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1428337_at | - | - |
Mouse Genome 430 2.0 Array | 1450067_a_at | - | - |
Mouse Genome 430 2.0 Array | 1420974_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67881_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67881_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1400016056.mm.1 | TC1400002093.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016056.mm.1 | NM_023397.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016056.mm.1 | NM_023397.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016056.mm.1 | NR_028316.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016056.mm.1 | NR_028316.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016057.mm.1 | TC1400002093.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1400016057.mm.1 | NM_023397.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016057.mm.1 | NM_023397.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016057.mm.1 | NR_028316.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1400016057.mm.1 | NR_028316.1 | Refseq |
Mouse Expression Set 430 | 1450067_PM_a_at | - | - |
Mouse Expression Set 430 | 1420974_PM_at | - | - |
Mouse Expression Set 430 | 1420974_at | - | - |
Mouse Expression Set 430 | 1428337_at | - | - |
Mouse Expression Set 430 | 1450067_a_at | - | - |
Mouse Expression Set 430 | 1428337_PM_at | - | - |
Murine Genome U74v2 Set | 95447_PM_at | - | - |
Murine Genome U74v2 Set | 95447_at | - | - |
Mouse Genome 430A 2.0 Array | 1428337_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1420974_at | - | - |
Mouse Genome 430A 2.0 Array | 1450067_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1450067_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1420974_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1428337_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428337_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450067_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428337_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450067_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 672707 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 106100021 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 940465 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000940465 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244756 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1239137 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244756 | - | - |
mouse-6 v1.1 (Illumina) | scl45535.5_307-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106100021 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 940465 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl45535.5_307-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106100021 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940465 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244756 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244756 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1239137 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl45535.5_307-S | - | - |
