Assay Details
Gene Symbol: Slc44a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023557.3 | NP_076046.1 | 4 - 5 | 260 | 99 | |
GenBank mRNA | AK145621.1 | - | 4 - 5 | 252 | 99 | |
BC010808.1 | - | 4 - 5 | 258 | 4240658 | 99 | |
BC052652.1 | - | 4 - 5 | 258 | 30034507 | 99 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
