Assay Details
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 2 - 3 | 217 | 123 | |
GenBank mRNA | AK004594.2 | - | 2 - 3 | 191 | 123 | |
AK013437.1 | - | 2 - 3 | 195 | 123 | ||
AK160038.1 | - | 2 - 3 | 187 | 123 | ||
BC003278.1 | - | 2 - 3 | 146 | 3582180 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK160038.1, BC003278.1, NM_024258.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
