Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 5 - 6 | 1109 | 95 | ||
GenBank mRNA | AK006249.1 | - | 3 - 4 | 375 | 95 | |
AK016463.1 | - | 5 - 6 | 1121 | 95 | ||
AY461596.1 | - | 5 - 6 | 1127 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
