Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 7 - 8 | 1359 | 132 | ||
GenBank mRNA | AK006249.1 | - | 5 - 6 | 625 | 132 | |
AK016463.1 | - | 7 - 8 | 1371 | 132 | ||
AY461596.1 | - | 7 - 8 | 1377 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
