Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 9 - 10 | 1705 | 89 | ||
GenBank mRNA | AK006249.1 | - | 7 - 8 | 971 | 89 | |
AY461596.1 | - | 9 - 10 | 1723 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
