Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028119.5 | NP_082395.2 | 1 - 2 | 374 | 64 | |
GenBank mRNA | AK011756.1 | - | 1 - 2 | 217 | 64 | |
AK158770.1 | - | 1 - 2 | 254 | 64 | ||
AY027937.1 | - | 1 - 2 | 292 | 64 | ||
BC028955.1 | - | 1 - 2 | 43 | 64 | ||
BC080821.1 | - | 1 - 2 | 195 | 64 | ||
BC099410.1 | - | 1 - 2 | 141 | 30933658 | 64 | |
CK627126.1 | - | 1 - 2 | 374 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
